Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APCDD1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11092756UV
Description
APCDD1 Polyclonal specifically detects APCDD1 in Human, Mouse, Chicken, Reptile samples. It is validated for Western Blot.Specifications
APCDD1 | |
Polyclonal | |
Western Blot | |
adenomatosis polyposis coli down-regulated 1, Adenomatosis polyposis coli down-regulated 1 protein, B7323, DRAPC1, protein APCDD1 | |
Rabbit | |
Affinity Purified | |
Cancer | |
147495.0 | |
Store at 4°C in the dark. |
KnockDown | |
DyLight 350 | |
50 mM sodium borate with 0.05% sodium azide | |
APCDD1 | |
Synthetic peptide made to an internal portion of human APCDD1 (within residues 400-500). [Swiss-ProtNo. Q8J025] | |
0.1 mL | |
Primary | |
Human, Mouse, Chicken, Reptile | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction