Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APCDD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598605 Supplier Invitrogen™ Supplier No. PA598605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APCDD1 Polyclonal Antibody for ELISA, IHC (P), Western Blot

APCDD1 (Drapc1) is a novel protein that has been shown to be a target of Wnt/beta-catenin signaling pathway in cancer cell lines. APCDD1 is overexpressed in colorectal carcinogenesis and is deregulated in CTNNB1-mutated Wilms tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APCDD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene APCDD1
Gene Accession No. Q8J025
Gene Alias AB023957; adenomatosis polyposis coli down-regulated 1; adenomatosis polyposis coli down-regulated 1 protein; Adenomatosis polyposis coli down-regulated 1 protein homolog; APC down-regulated 1; apcdd1; APCDD1 (HGNC:15718); APCDD1 homologue; AU041258; B7323; cAPCDD1; Drapc1; EIG180; ethanol induced gene product EIG180; FP7019; HHS; HTS; hypoptrichosis simplex; HYPT1; Pgo1; primglo1; protein APCDD1; Protein primglo1
Gene Symbols APCDD1
Host Species Rabbit
Immunogen Recombinant Human Protein APCDD1 protein (22-288AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147495
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.