Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APCDD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572925
Description
APCDD1 Polyclonal Antibody for Western Blot
APCDD1 (Drapc1) is a novel protein that has been shown to be a target of Wnt/beta-catenin signaling pathway in cancer cell lines. APCDD1 is overexpressed in colorectal carcinogenesis and is deregulated in CTNNB1-mutated Wilms tumors.
Specifications
APCDD1 | |
Polyclonal | |
Unconjugated | |
APCDD1 | |
AB023957; adenomatosis polyposis coli down-regulated 1; adenomatosis polyposis coli down-regulated 1 protein; Adenomatosis polyposis coli down-regulated 1 protein homolog; APC down-regulated 1; apcdd1; APCDD1 (HGNC:15718); APCDD1 homologue; AU041258; B7323; cAPCDD1; Drapc1; EIG180; ethanol induced gene product EIG180; FP7019; HHS; HTS; hypoptrichosis simplex; HYPT1; Pgo1; primglo1; protein APCDD1; Protein primglo1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147495, 426765, 494504 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.3 mg/mL | |
PBS with 30% glycerol and 0.1% sodium azide | |
Q3U128, Q5R2I8, Q8J025 | |
APCDD1 | |
Synthetic peptide made to an internal portion of human APCDD1 (within residues 400-500). [Swiss-Prot# Q8J025]. | |
100 μL | |
Primary | |
Human, Mouse, Chicken, Reptile | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction