Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human APCDD1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | APCDD1 |
---|---|
Clone | 2584B |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10501SP
![]() |
R&D Systems
MAB10501SP |
25 μg |
Each for $200.50
|
|
|||||
MAB10501100
![]() |
R&D Systems
MAB10501100 |
100 μg |
Each for $502.50
|
|
|||||
Description
APCDD1 Monoclonal specifically detects APCDD1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
APCDD1 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
adenomatosis polyposis coli down-regulated 1, Adenomatosis polyposis coli down-regulated 1 protein, APC Down Regulated 1, B7323, DRAPC1, HYPT1, protein APCDD1 | |
Mouse myeloma cell line, NS0-derived human APCDD1, Leu27-His492, Accession # Q8J025 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2584B | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Rabbit | |
Human | |
Q8J025 | |
147495 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title