Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APCDD1L Antibody (Z35P1F10*C4), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250107AF750
Description
APCDD1L Monoclonal specifically detects APCDD1L in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
APCDD1L | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
APCDD1L | |
Peptide Sequence -KRPTSYQA (immunogen peptide is identical in sheep, bovine, porcine, equine and chicken) | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
Z35P1F10*C4 | |
Western Blot, ELISA, Immunohistochemistry | |
Adenomatosis polyposis coli down-regulated 1 protein-like, adenomatosis polyposis coli down-regulated 1-like, FLJ90166, MGC126807, MGC126809, protein APCDD1-like, RP4-685L9.2 | |
Mouse | |
Protein A purified | |
Cellular Signaling | |
164284 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction