Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APE Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 64 publications
$243.50 - $501.50
Specifications
Antigen | APE |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1:1000, Simple Western 1:12.5, Flow Cytometry, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 7 ug/ml, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Immunohistochemistry Free-Floating reported in scientific literature (PMID 17332344), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Block/Neutralize |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100101SS
![]() |
Novus Biologicals
NB100101SS |
0.025 mL |
Each for $243.50
|
|
|||||
NB100101
![]() |
Novus Biologicals
NB100101 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
APE Polyclonal specifically detects APE in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Block/Neutralize.Specifications
APE | |
Western Blot 1:1000, Simple Western 1:12.5, Flow Cytometry, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 7 ug/ml, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Immunohistochemistry Free-Floating reported in scientific literature (PMID 17332344), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Block/Neutralize | |
Polyclonal | |
Rabbit | |
Autophagy, Base Excision Repair, Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease | |
APEX1 | |
IgG | |
Affinity Purified | |
Human, mouse, primate and rat. Rabbit reactivity reported in the scientific literature (PMID: 15276530). |
0.5 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Primate, Rabbit | |
P27695 | |
328 | |
Affinity purified human APE1 [UniProt# P27695] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
37 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title