Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APEH Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB060209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB060209 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB060209 Supplier Novus Biologicals Supplier No. NBP299357JF549
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APEH Polyclonal specifically detects APEH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen APEH
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AARE, ACPH, acylamino-acid-releasing enzyme, acylaminoacyl-peptidase, Acyl-peptide hydrolase, APH, D3F15S2, D3S48EEC 3.4.19.1, DNF15S2, MGC2178, N-acylaminoacyl-peptide hydrolase, OPH, Oxidized protein hydrolase
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human APEH.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 327
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.