Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APEH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543138
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543138 Supplier Invitrogen™ Supplier No. PA543138
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GSTGFGQDSI LSLPGNVGHQ DVKDVQFAVE QVLQEEHFDA SHVALMGGSH Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 92%.

APEH is the enzyme acylpeptide hydrolase, which catalyzes the hydrolysis of the terminal acetylated amino acid preferentially from small acetylated peptides. The acetyl amino acid formed by this hydrolase is further processed to acetate and a free amino acid by an aminoacylase. APEH gene is located within the same region of chromosome 3 (3p21) as the aminoacylase gene, and deletions at this locus are also associated with a decrease in aminoacylase activity. The acylpeptide hydrolase is a homotetrameric protein of 300 kDa with each subunit consisting of 732 amino acid residues. The protein can play an important role in destroying oxidatively damaged proteins in living cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APEH
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene APEH
Gene Accession No. P13798
Gene Alias AARE; ACPH; Acylamino-acid-releasing enzyme; acylamino-acid-releasing enzyme-like; acylaminoacyl-peptidase; acylaminoacyl-peptide hydrolase; acylpeptide hydrolase; Acyl-peptide hydrolase; Apeh; APH; cb5; D3F15S2; D3S48E; DNF15S2; LOW QUALITY PROTEIN: acylamino-acid-releasing enzyme; acylamino-acid-releasing enzyme; MGC2178; N-acylaminoacyl peptide hydrolase; N-acylaminoacyl-peptide hydrolase; OPH; Oxidized protein hydrolase; sb:cb5; wu:fi37d02
Gene Symbols APEH
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human APEH.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 327
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.