Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APEH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597467
Description
APEH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
APEH is the enzyme acylpeptide hydrolase, which catalyzes the hydrolysis of the terminal acetylated amino acid preferentially from small acetylated peptides. The acetyl amino acid formed by this hydrolase is further processed to acetate and a free amino acid by an aminoacylase. APEH gene is located within the same region of chromosome 3 (3p21) as the aminoacylase gene, and deletions at this locus are also associated with a decrease in aminoacylase activity. The acylpeptide hydrolase is a homotetrameric protein of 300 kDa with each subunit consisting of 732 amino acid residues. The protein can play an important role in destroying oxidatively damaged proteins in living cells.
Specifications
APEH | |
Polyclonal | |
Unconjugated | |
APEH | |
AARE; ACPH; Acylamino-acid-releasing enzyme; acylamino-acid-releasing enzyme-like; acylaminoacyl-peptidase; acylaminoacyl-peptide hydrolase; acylpeptide hydrolase; Acyl-peptide hydrolase; Apeh; APH; cb5; D3F15S2; D3S48E; DNF15S2; LOW QUALITY PROTEIN: acylamino-acid-releasing enzyme; acylamino-acid-releasing enzyme; MGC2178; N-acylaminoacyl peptide hydrolase; N-acylaminoacyl-peptide hydrolase; OPH; Oxidized protein hydrolase; sb:cb5; wu:fi37d02 | |
Rabbit | |
Protein G | |
RUO | |
327 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P13798 | |
APEH | |
Recombinant Human Acylamino-acid-releasing enzyme protein (201-500AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction