Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APEH Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5110984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110984 Supplier Thermo Scientific Supplier No. PA5110984

Rabbit Polyclonal Antibody

Immunogen sequence: IARLKKPDQA IKGDQFVFYE DWGENMVSKS IPVLCVLDVE SGNISVLEGV PENVSPGQAF WAPGDAGVVF VGWWHEPFRL GIRFCTNRRS A

APEH is the enzyme acylpeptide hydrolase, which catalyzes the hydrolysis of the terminal acetylated amino acid preferentially from small acetylated peptides. The acetyl amino acid formed by this hydrolase is further processed to acetate and a free amino acid by an aminoacylase. APEH gene is located within the same region of chromosome 3 (3p21) as the aminoacylase gene, and deletions at this locus are also associated with a decrease in aminoacylase activity. The acylpeptide hydrolase is a homotetrameric protein of 300 kDa with each subunit consisting of 732 amino acid residues. The protein can play an important role in destroying oxidatively damaged proteins in living cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APEH
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APEH
Gene Accession No. P13798
Gene Alias AARE; ACPH; Acylamino-acid-releasing enzyme; Acylaminoacyl-peptidase; Acyl-peptide hydrolase; Apeh; APH; BOS_20965; cb5; D3F15S2; D3S48E; DNF15S2; MGC2178; OPH; Oxidized protein hydrolase; sb:cb5; wu:fi37d02
Gene Symbols APEH
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human APEH
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 327
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.