Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apelin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP231176
Description
Apelin Polyclonal specifically detects Apelin in Human, Mouse, Porcine, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Apelin | |
Polyclonal | |
Unconjugated | |
Tris 0,1M, glycine 0,1M, sucrose 2% with No Preservative | |
APLN | |
Synthetic peptide derived from C terminal region of human Apelin | |
0.1 mg | |
Primary | |
Reacts with a human. Predicted to cross react with rat, mouse and bovine based upon sequence homology. | |
Human, Mouse, Sheep | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
LYOPH | |
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin | |
AGTRL1 ligand, APEL, apelin, apelin, AGTRL1 ligand, APJ endogenous ligand, XNPEP2 | |
Rabbit | |
Protein A purified | |
RUO | |
8862 | |
Lyophilized powder. Reconstitute in 100ul distilled water. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction