Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Apelin Receptor/APJ, Mouse, Unlabeled, Clone: 3C3-7,

Catalog No. MABN1846MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1846MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1846MI Supplier MilliporeSigma™ Supplier No. MABN1846
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apelin receptor (UniProt P35414; also known as Angiotensin receptor-like 1, G-protein coupled receptor APJ, G-protein coupled receptor HG11) is encoded by the APLNR (also known as AGTRL1, APJ) gene (Gene ID 187) in human. Apelin is an angiotensin converting enzyme 2 (ACE2) substrate that functions as an inotropic and cardiovascular protective peptide. Apelin exerts its physiological effects via its receptor APJ, a seven-transmembrane G protein-coupled receptor that shares significant homology with the Ang II type 1 receptor (AT1R). Apelin/APJ signaling plays an important role in a wide range of pathophysiological effects in the cardiovascular system. Apelin levels are reduced in patients with essential hypertension independent of left ventricular systolic and diastolic dysfunction. Treatment with Apelin has been shown to lower systolic blood pressure levels in hypertensive rats via an NO-dependent mechanism. APJ is a 7-transmembrane (a.a. 27-51, 67-91, 101-125, 145-166, 201-221, 245-271, 285-308) GPCR, having a short N-terminal extracellular portion (a.a. 1-26) and a long C-terminal cytomplasmic tail (a.a. 309-380).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apelin Receptor/APJ
Applications Western Blot
Classification Monoclonal
Clone 3C3-7
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene APLNR, AGTRL1, APJ
Gene Accession No. P35414
Host Species Mouse
Immunogen Recombinant full-length human Apelin receptor/APJ.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_005152
Test Specificity Clone 3C3-7 detects human Apelin receptor/APJ.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.