Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apg12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985834
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5834 Supplier Bioss Supplier No. BS4012R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ATG12, a member of the autophagy protein family, forms a conjugate with ATG5; this conjugate has a ubiquitin-protein ligase (E3)-like activity for protein lipidation in autophagy. This conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells. ATG12 has also been shown to interact with ATG10 in human embryonic kidney cells in the presence of ATG7. At least two isoforms of ATG12 are known to exist. Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1.

Specifications

Antigen Apg12
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ATG12
Gene Accession No. O94817
Gene Alias 4931423H11Rik; A330058M13Rik; APG 12; Apg12; Apg12 (autophagy, yeast) homolog; Apg12l; APG12-like; ATG12; ATG12 autophagy related 12 homolog; Atg12l; Atg12-PB; Autophagy protein 12-like; autophagy related 12; autophagy related protein 12; Autophagy-related 12; Autophagy-related protein 12; Autophagy-specific gene 12; CG10861; CG10861 gene product from transcript CG10861-RB; CG10861-PB; Dmel\CG10861; Dmel_CG10861; FBR93; HAPG12; OTTHUMP00000159129; Ubiquitin-like protein ATG12
Gene Symbols ATG12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Apg12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9140
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.