Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ APH-1A, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABC1696MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABC1696MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC1696MI Supplier MilliporeSigma™ Supplier No. ABC1696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gamma-secretase subunit APH-1A (UniProt Q96BI3; also known as Anterior pharynx defective 1 homolog A, APH-1a, Aph-1alpha, Presenilin-stabilization factor) is encoded by the APH1A (also known as PSF, CGI-78) gene (ORF UNQ579/PRO1141; Gene ID 51107) in human. Gamma-secretase catalyzes the final cleavage of beta-amyloid precursor protein to generate amyloid-beta peptide, the principal component of amyloid plaques found in Alzheimer's diseased (AD) brains. Gamma-secretase is a complex composed of four integral membrane proteins, presenilin (PS), nicastrin (NCT), PEN-2, and APH-1. The PS component could be either a PS1 or PS2 and the APH component could be either an APH-1a or APH-1b, alternative splicings of these four proteins contribute to further heterogeneity among gamma-secretase complexes. The gamma-secretase complex is assembled in the ER, where NCT and APH-1 form an initial scaffold for the PS holoprotein, which becomes stabilized upon interaction with the NCT/APH-1. Subsequent addition of PEN-2 to this trimeric assembly intermediate triggers the endoproteolytic cleavage of the PS holoprotein. The fourth transmembrane domain (TMD4) of APH-1, the PEN-2 C-terminal end, the NCT TMD domain, and the PS1 C-terminal end are shown to be essential for functional gamma-secretase complex assembly. APH-1a is a seven-transmembrane (a.a. 3-23, 32-52, 69-89, 119-139, 159-179, 187-207, 214-234) protein with a lumenal N-terminus and a cytoplasmic C-terminal tail (a.a. 235-265).
TRUSTED_SUSTAINABILITY

Specifications

Antigen APH-1A
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene APH1A, PSF, CGI-78, UNQ579/PRO1141
Gene Accession No. Q96BI3
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence from the C-terminal cytoplasmic tail of human APH-1A.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001071096
Test Specificity Expected to react with all three spliced isoforms of human APH-1A reported by UniProt (Q96BI3).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.