Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aph 1b Rabbit anti-C. elegans, Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206081
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6081 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6081 Supplier Fabgennix Inc Supplier No. APH1B101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aph 1b Polyclonal antibody specifically detects Aph 1b in C. elegans, Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Aph 1b
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene APH1B
Gene Accession No. Q8C7N7, Q8WW43
Gene Alias 2310057K14Rik; anterior pharynx defective 1 homolog B; anterior pharynx defective 1b homolog; anterior pharynx defective 1b homolog (C. elegans); anterior pharynx defective 1c homolog; aph-1 homolog B, gamma secretase subunit; aph-1 homolog B, gamma-secretase subunit; Aph1b; APH-1b; APH-1B'; APH1B gamma secretase subunit; APH1B homolog; aph-1beta; Aph1c; Gamma-secretase subunit APH-1B; LOW QUALITY PROTEIN: gamma-secretase subunit APH-1B; Presenilin-stabilization factor-like; PRO1328; PSFL; TAAV688; UNQ688/PRO1328
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 235-257 of Human Presenilin stabilizing protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117883761, 208117, 300802, 83464
Target Species C. elegans, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.