Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aph-1b Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981541
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1541 Supplier Bioss Supplier No. BS11640R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APH1B gene ontology annotations related to this gene include Notch receptor processing; Notch signaling pathway; positive regulation of catalytic activity; protein processing.

Specifications

Antigen Aph-1b
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene APH1B
Gene Accession No. Q8WW43
Gene Alias 2310057K14Rik; anterior pharynx defective 1 homolog B; anterior pharynx defective 1 homolog B (C. elegans); anterior pharynx defective 1b homolog; anterior pharynx defective 1b homolog (C. elegans); anterior pharynx defective 1c homolog; aph-1 homolog B, gamma secretase subunit; aph-1 homolog B, gamma-secretase subunit; Aph1b; APH-1b; APH-1B′; APH1B gamma secretase subunit; APH1B homolog; Aph-1beta; Aph1c; BOS_10822; Gamma-secretase subunit APH-1B; Presenilin-stabilization factor-like; PRO1328; PSFL; TAAV688; UNQ688/PRO1328
Gene Symbols APH1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Aph-1b.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83464
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.