Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APH1a Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986018
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6018 Supplier Bioss Supplier No. BS4259R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APH1 is an essential subunit of the gamma-secretase complex, an endoprotease complex that catalyzes the intramembrane cleavage of integral proteins such as Notch receptors and APP (beta-amyloid precursor protein). The gamma-secretase complex is minimally composed of a presenilin homodimer (PSEN1 or PSEN2), nicastrin (NCSTN), APH1 (APH1A or APH1B) and PEN2, although other components may exist. APH1 probably represents a stabilizing cofactor for the presenilin homodimer that promotes the formation of a stable complex.

Specifications

Antigen APH1a
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene APH1A
Gene Accession No. Q96BI3
Gene Alias 6530402N02Rik; anterior pharynx defective 1 homolog A; anterior pharynx defective 1 homolog A (C. elegans); anterior pharynx defective 1a homolog; anterior pharynx defective 1a homolog (C. elegans); APH1; APH-1; aph-1 homolog A, gamma secretase subunit; aph-1 homolog A, gamma-secretase subunit; APH1A; APH-1a; APH1A gamma secretase subunit; Aph-1alpha; BOS_2946; CGI-78; Gamma-secretase subunit APH-1A; Presenilin-stabilization factor; PSF; RGD1311546; RP4-790G17.3; UNQ579/PRO1141
Gene Symbols APH1A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human APH1a.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51107
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.