Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Aphidicolin

Catalog No. ac611970010
Click to view available options
Quantity:
1 mg
5 mg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38966-21-1
Molecular Formula C20H34O4
Molecular Weight (g/mol) 338.488
MDL Number MFCD00083214
InChI Key NOFOAYPPHIUXJR-APNQCZIXSA-N
Synonym aphidicolin, unii-192tj6pp19, ccris 1783, chembl29711, 3r,4r,4ar,6as,8r,9r,11as,11bs-4,9-bis hydroxymethyl-4,11b-dimethyltetradecahydro-8,11a-methanocyclohepta a naphthalene-3,9-diol, 9,15-cyclo-c,18-dinor-14,15-secoandrostane-4,17-dimethanol, 3,17-dihydroxy-4-methyl-, 3alpha,4alpha,5alpha,17alpha, +-aphidicolin, 8,11a-methano-11ah-cyclohepta a naphthalene-4,9-dimethanol, tetradecahydro-3,9-dihydroxy-4,11b-dimethyl-, 3r-3-alpha,4-alpha,4a-alpha,6a-beta,8-beta,9-beta,11a-beta,11b-beta, +/--aphidicolin, aphidicolin, +/-
PubChem CID 457964
ChEBI CHEBI:2766
SMILES CC12CCC(C(C1CCC3C24CCC(C(C3)C4)(CO)O)(C)CO)O

Specifications

Melting Point 226°C to 235°C
Quantity 1 mg
Formula Weight 338.49
Percent Purity 98% min.
Chemical Name or Material Aphidicolin
RTECSNumber PB9185000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.