Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ API5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555354
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555354 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555354 Supplier Invitrogen™ Supplier No. PA555354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVEELYRNYG ILADATEQVG QHKDAYQVIL DGVKGGTKEK RLAAQFIPKF FKHFPELADS AINAQLDLCE DEDVSIRRQA IKELPQFATG ENLPRVADIL TQLLQTDDSA EFNLVN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes an apoptosis inhibitory protein whose expression prevents apoptosis after growth factor deprivation. This protein suppresses the transcription factor E2F1-induced apoptosis and also interacts with, and negatively regulates Acinus, a nuclear factor involved in apoptotic DNA fragmentation. Its depletion enhances the cytotoxic action of the chemotherapeutic drugs. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen API5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene API5
Gene Accession No. O35841, Q9BZZ5
Gene Alias AAC11; AAC-11; AI196452; antiapoptosis clone 11 protein; API5; API-5; Apoptosis inhibitor 5; cell migration-inducing gene 8 protein; Fibroblast growth factor 2-interacting factor; fibroblast growth factor 2-interacting factor 2; FIF; FIF C1; MIG8; migration-inducing protein MIG8; Protein XAGL
Gene Symbols API5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human API5. Recombinant protein control fragment (Product #RP-92069).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11800, 362170, 8539
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.