Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apigenin, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Protein kinase inhibitor. Suppresses tumor-promoting effects of TPA and exhibits antiproliferative activity in human breast cancer cells (IC50 values are 59.44 and 31.15 μM at 24 and 72 hrs respectively). Activates both the intrinsic and extrinsic apoptotic pathways and displays anti-inflammatory, hypotensive, antispasmodic and antioxidant properties in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 270.2 |
| Percent Purity | >98% |
| Chemical Name or Material | Apigenin |
Safety and Handling
| Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction