Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APIP Antibody (19F461), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056567H
Description
APIP Monoclonal specifically detects APIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
APIP | |
Monoclonal | |
HRP | |
Q96GX9 | |
APIP | |
The antibody was developed against a full-length His-tagged recombinant APIP. | |
0.1 mL | |
Apoptosis | |
51074 | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
19F461 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
APAF1 interacting protein, APAF1-interacting protein, APIP2, CGI29, CGI-29, dJ179L10.2, EC 4.2.1.109, MMRP19, MTRu-1-P dehydratase, probable methylthioribulose-1-phosphate dehydratase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This monoclonal antibody can be used for detection of APIP and APIP2. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction