Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APIP Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen APIP
Concentration 0.5 mg/mL
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056383T
SDP
View Documents
Novus Biologicals
NB10056383SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NB10056383
SDP
View Documents
Novus Biologicals
NB10056383
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

APIP Polyclonal specifically detects APIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

APIP
Western Blot 1-2 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human
Q96GX9
51074
The antibody was developed against a full-length His-tagged recombinant APIP protein (NP_057041).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis
PBS containing 0.2% gelatin with 0.05% Sodium Azide
APAF1 interacting protein, APAF1-interacting protein, APIP2, CGI29, CGI-29, dJ179L10.2, EC 4.2.1.109, MMRP19, MTRu-1-P dehydratase, probable methylthioribulose-1-phosphate dehydratase
APIP
IgG
Protein G purified
This polyclonal can be used for detection of APIP and APIP2.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.