Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582749 Supplier Invitrogen™ Supplier No. PA582749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSGCDAREGD CCSRRCGAQD KEHPRYLIPE LCKQFYHLGW VTGTGGGISL KHGDEIYIAP SGVQKERIQP EDMFVC Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 92%.

The mammalian homologues of the key cell death gene CED-4 in C. elegans has been identified recently from human and mouse and designated Apaf 1 (for apoptosis protease-activating factor 1). Apaf 1 binds to cytochrome c (Apaf 2) and Caspase-9 (Apaf 3), which leads to Caspase-9 activation. Activated Caspase-9 in turn cleaves and activates Caspase-3 that is one of the key proteases, being responsible for the proteolytic cleavage of many key proteins in apoptosis. Recently, Cho et al (4) have identified a new Apaf 1 Interactiong Protein (APIP) also known as CG129 and MMRP19, as a negative regulator of ischemic injury. APIP competes with Caspase-9 binding site of Apaf 1. APIP is predicted to code for a 204 amino acid. An isoform of APIP, APIP2 encodes a 242 amino acid protein, which is an alternative splicing variant differing in its N-terminus from APIP. APIP transcript is ubiquitously expressed in most adult tissue with high expression in skeletal muscle, heart, and kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APIP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APIP
Gene Accession No. Q96GX9
Gene Alias APAF1 interacting protein; APAF1-interacting protein; APIP; APIP2; CG129; CGI29; CGI-29; dJ179L10.2; hAPIP; Methylthioribulose-1-phosphate dehydratase; Mmrp19; monocyte macrophage 19; monocyte/macrophage protein 19; MTRu-1-P dehydratase; probable methylthioribulose-1-phosphate dehydratase; RGD1564562
Gene Symbols APIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APIP. Recombinant protein control fragment (Product #RP-92053).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.