Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APLNRA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572239
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572239 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572239 Supplier Invitrogen™ Supplier No. PA572239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APLNRA Polyclonal Antibody for Western Blot

Receptor for apelin coupled to G proteins that inhibit adenylate cyclase activity and plays a role in various processes in adults such as regulation of blood pressure, heart contractility, and heart failure. Also plays a key role in early development such as gastrulation and heart morphogenesis by acting as a receptor for apela hormone, promoting endoderm and mesendoderm cell migration and regulating the migration of cells fated to become myocardial progenitors, respectively (PubMed:24316148, PubMed:24407481). Acts redundantly with agtrl1b in heart development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APLNRA
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene aplnra
Gene Accession No. Q7SZP9
Gene Alias Agtr1; agtrl1; agtrl1a; angiotensin II receptor-like 1a; angiotensin receptor-like 1; angiotensin receptor-like 1a; Apelin receptor A; aplnra; G-protein coupled receptor APJ A; msr; zgc:152843
Gene Symbols aplnra
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 343-377 amino acids from the human region of DANRE aplnra.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 561935
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.