Learn More
Invitrogen™ APLP1 Monoclonal Antibody (354008)

Description
In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse APLP-1 is observed and no cross-reactivity with recombinant human APLP-2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | APLP1 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 354008 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | APLP1 |
| Gene Accession No. | P51693 |
| Gene Alias | amyloid beta (A4) precursor-like protein 1; amyloid beta precursor like protein 1; amyloid precursor-like protein 1; amyloid-like protein 1; APLP; APLP 1; APLP1; APLP-1; Aplp1_retired; APLPAPLP-1; C30; RGD1561211 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.