Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APLP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120825 Supplier Invitrogen™ Supplier No. PA5120825
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: C6, Mouse brain, Rat brain. The target is usually found in the following locations: Cell membrane, Cytoplasm, Single-pass type I membrane protein. Immunogen sequence: NPHLAQELRP QIQELLHSEH LGPSELEAPA PGGSSEDKGG LQPPDSKDDT PMTLPKGSTE QDAASPEKEK MNPLEQYERK VNASVPRGFP FHSSEIQRDE LAPAGTGVSR E.

This gene encodes a member of the highly conserved amyloid precursor protein gene family. The encoded protein is a membrane-associated glycoprotein that is cleaved by secretases in a manner similar to amyloid beta A4 precursor protein cleavage. This cleavage liberates an intracellular cytoplasmic fragment that may act as a transcriptional activator. The encoded protein may also play a role in synaptic maturation during cortical development. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APLP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene APLP1
Gene Accession No. P51693, Q03157
Gene Alias amyloid beta (A4) precursor-like protein 1; amyloid beta precursor like protein 1; amyloid precursor-like protein 1; amyloid-like protein 1; APLP; APLP 1; APLP1; APLP-1; Aplp1_retired; APLPAPLP-1; C30; RGD1561211
Gene Symbols APLP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 470-580 of human APLP1 (NP_0051571).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11803, 333, 502317
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.