Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APLP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556020 Supplier Invitrogen™ Supplier No. PA556020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLAQELRPQI QELLHSEHLG PSELEAPAPG GSSEDKGGLQ PPDSKDDTPM TLPKGSTEQD AASPEKEKMN PLEQYERKVN ASVPRGFPFH SSEIQRDELA PAGTGVSLGA PHR Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 70%.

This gene encodes a member of the highly conserved amyloid precursor protein gene family. The encoded protein is a membrane-associated glycoprotein that is cleaved by secretases in a manner similar to amyloid beta A4 precursor protein cleavage. This cleavage liberates an intracellular cytoplasmic fragment that may act as a transcriptional activator. The encoded protein may also play a role in synaptic maturation during cortical development. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APLP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APLP1
Gene Accession No. P51693
Gene Alias amyloid beta (A4) precursor-like protein 1; amyloid beta precursor like protein 1; amyloid precursor-like protein 1; amyloid-like protein 1; APLP; APLP 1; APLP1; APLP-1; Aplp1_retired; APLPAPLP-1; C30; RGD1561211
Gene Symbols APLP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APLP1. Recombinant protein control fragment (Product #RP-102552).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 333
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.