Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APLP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 150411AP150UL

Description
APLP2 May play a role in the regulation of hemostasis. The soluble form may have inhibitory properties towards coagulation factors. May interact with cellular G-protein signaling pathways. May bind to the DNA 5′-GTCACATG-3′ (CDEI box).Specifications
APLP2 | |
Polyclonal | |
Unconjugated | |
APLP2 | |
Amyloid like protein 2, Amyloid protein homolog, APLP 2, APLP2, APPH, APPL2, CDEBP, CDEI box binding protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11804, 334, 64312 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15943, Q06335, Q06481 | |
APLP2 | |
APLP2 Fusion Protein Ag6838 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction