Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555424
Description
Immunogen sequence: LKQQVEGTAQ EAVSAAGAAA QQVVDQATEA GQKAMDQLAK TTQETID Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 34%.
APM2 gene is exclusively expressed in adipose tissue. Its function is currently unknown.
Specifications
| APM2 | |
| Polyclonal | |
| Unconjugated | |
| ADIRF | |
| Adipogenesis factor rich in obesity; adipogenesis regulatory factor; Adipose most abundant gene transcript 2 protein; adipose specific 2; adipose-specific protein 2; ADIRF; AFRO; APM2; apM-2; C10orf116; RP11-96C23.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10974 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.08 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15847 | |
| ADIRF | |
| Recombinant protein corresponding to Human APM2. Recombinant protein control fragment (Product #RP-88971). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction