Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA112909
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA112909 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA112909 Supplier Invitrogen™ Supplier No. PA112909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control for this product is adipose tissue, cornea or liver tissue.

APM2 gene is exclusively expressed in adipose tissue. Its function is currently unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APM2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene ADIRF
Gene Accession No. Q15847
Gene Alias Adipogenesis factor rich in obesity; adipogenesis regulatory factor; Adipose most abundant gene transcript 2 protein; adipose specific 2; adipose-specific protein 2; ADIRF; AFRO; APM2; apM-2; C10orf116; RP11-96C23.4
Gene Symbols ADIRF
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human APM2 protein.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10974
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.