Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APMAP Antibody (OTI4F6), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272080FR
Description
APMAP Monoclonal specifically detects APMAP in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
APMAP | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
APMAP | |
Full length recombinant protein of human APMAP (NP_065392) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI4F6 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
adipocyte plasma membrane-associated protein, APMAP, BSCv, chromosome 20 open reading frame 3, Protein BSCv | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
57136.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction