Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Apo-ONE™ Homogeneous Caspase-3/7 Buffer

Catalog No. PRG7781 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG7781 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG7781 Supplier Promega Supplier No. G7781
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Component for measuring active Caspase-3 and -7 in a homogeneous format.

Apo-ONE™ Homogeneous Caspase-3/7 Buffer is a component of the Apo-ONE™ Homogeneous Caspase-3/7 Assay (Mfr. Nos. G7792, G7790 and G7791).

Specifications

Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.