Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOA1 Mouse anti-Human, Mouse, Clone: 464CT10.4.4, Abnova™

Catalog No. 89933574 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-574 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-574 Supplier Abnova Corporation Supplier No. MAB12867

Mouse monoclonal antibody raised against synthetic peptide of human APOA1.

This gene encodes apolipoprotein A-I, which is the major protein component of high density lipoprotein (HDL) in plasma. The protein promotes cholesterol efflux from tissues to the liver for excretion, and it is a cofactor for lecithin cholesterolacyltransferase (LCAT) which is responsible for the formation of most plasma cholesteryl esters. This gene is closely linked with two other apolipoprotein genes on chromosome 11. Defects in this gene are associated with HDL deficiencies, including Tangier disease, and with systemic non-neuropathic amyloidosis. [provided by RefSeq]

Specifications

Antigen APOA1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 464CT10.4.4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human APOA1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:100-1:800) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.09% sodium azide).
Gene Alias MGC117399
Gene Symbols APOA1
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus region of human APOA1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 335
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.