Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ApoA1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142393
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
Specifications
| ApoA1 | |
| Polyclonal | |
| Unconjugated | |
| APOA1 | |
| 1-Sep; 2-Sep; Alp-1; Apo a; APO A1; apo A-1; APO I; apo(a); APOA1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; apolipoprotein A-I; apolipoprotein A-I precursor; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; prepro-apolipoprotein-AI; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 335 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P02647 | |
| APOA1 | |
| Peptide with sequence C-SALEEYTKKLNTQ (aa 255-267). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction