Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980545
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0545 Supplier Bioss Supplier No. BS0849R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.

Specifications

Antigen APOA1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene APOA1
Gene Accession No. P02647
Gene Alias 1-Sep; 2-Sep; Alp-1; APO A1; apo A-1; APO I; apo(a); Apoa1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; Apolipoprotein A-I; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7
Gene Symbols APOA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human APOA1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 335
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.