Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ApoA1 Recombinant Superclonal™ Antibody (19HCLC)

Catalog No. PI710263
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI710263 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI710263 Supplier Invitrogen™ Supplier No. 710263
Add to Cart
Add to Cart

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with non-human primate and mouse based on sequence homology. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ApoA1
Applications Western Blot
Classification Recombinant Superclonal
Clone 19HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene APOA1
Gene Accession No. P02647, P04639
Gene Alias 1-Sep; 2-Sep; Alp-1; Apo a; APO A1; apo A-1; APO I; apo(a); APOA1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; apolipoprotein A-I; apolipoprotein A-I precursor; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; prepro-apolipoprotein-AI; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7
Gene Symbols APOA1
Host Species Rabbit
Immunogen Peptide corresponding to amino acids 127-142 of human Apo A1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25081, 335
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.