Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOA1BP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $648.50
Specifications
Antigen | APOA1BP |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
APOA1BP Polyclonal specifically detects APOA1BP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
APOA1BP | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
AIBPAI-BP, apolipoprotein A-I binding protein, apolipoprotein A-I-binding protein, MGC119143, MGC119144, MGC119145, YjeF N-terminal domain-containing protein 1, YjeF_N1, YJEFN1apoA-I binding protein | |
NAXE | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated | |
Polyclonal | |
Rabbit | |
Human | |
Q8NCW5 | |
128240 | |
This antibody was developed against a recombinant protein corresponding to amino acids: DIPSGWDVEKGNAGGIQPDLLISLTAPKKSATQFTGRYHYLGGRFVPPALEKKYQLNLPPYPDTECVYRL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title