Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ApoA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114033 Supplier Invitrogen™ Supplier No. PA5114033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKLLAATVLL LTICSLEGAL VRRQAKEPCV ESLVSQYFQT VTDYGKDLME For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 85%; Rabbit: 79%; Rat: 93%.

At least 9 distinct polymorphic forms of apolipoproteins are known. The apolipoproteins act as stabilizers of the intact lipoprotein particles. Quantitative measurements of HDL, LDL, and VLDL particles of human serum are often used to estimate an individual's relative risk of coronary heart disease. In addition, quantitative immunological measurements of certain apolipoproteins ( especially A-1 and B) have been suggested to be more accurate estimators of coronary heart disease that measurements of lipoprotein particles (especially HDL and LDL). The apolipoproteins are derived from human plasma by density gradient ultracentrifugation and HPLC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ApoA2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene APOA2
Gene Accession No. P02652
Gene Alias Alp-2; APO A2; APOA2; Apoa-2; ApoAII; Apo-AII; ApoA-II; Apolipoprotein; Apolipoprotein A2; Apolipoprotein AII; apolipoprotein A-II; Apolipoprotein A-II(1-76); Hdl-1; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-II
Gene Symbols APOA2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human APOA2 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 336
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.