Learn More
Invitrogen™ ApoA2 Recombinant Superclonal™ Antibody (43HCLC)
Rabbit Recombinant Superclonal Antibody
Supplier: Invitrogen™ 710260
Description
This antibody is predicted to react with non-human primate and mouse based on sequence homology. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
At least 9 distinct polymorphic forms of apolipoproteins are known. The apolipoproteins act as stabilizers of the intact lipoprotein particles. Quantitative measurements of HDL, LDL, and VLDL particles of human serum are often used to estimate an individual's relative risk of coronary heart disease. In addition, quantitative immunological measurements of certain apolipoproteins ( especially A-1 and B) have been suggested to be more accurate estimators of coronary heart disease that measurements of lipoprotein particles (especially HDL and LDL). The apolipoproteins are derived from human plasma by density gradient ultracentrifugation and HPLC.
Specifications
ApoA2 | |
Recombinant Superclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P02652, P04638 | |
APOA2 | |
Peptide corresponding to amino acids 60-71 of human Apo A2. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Western Blot | |
43HCLC | |
Unconjugated | |
APOA2 | |
Alp-2; APO A2; APOA2; Apoa-2; ApoAII; Apo-AII; ApoA-II; Apolipoprotein; Apolipoprotein A2; Apolipoprotein AII; apolipoprotein A-II; Apolipoprotein A-II(1-76); Hdl-1; ProapoA-II; Proapolipoprotein A-II; Truncated apolipoprotein A-II | |
Rabbit | |
Protein A | |
RUO | |
25649, 336 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.