Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOB Mouse anti-Human, Clone: F2C3, Abnova™

Catalog No. 89035399 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-399 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-399 Supplier Abnova Corporation Supplier No. MAB13994

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native human APOB.

  • High specificity
  • High reactivity

Specifications

Antigen APOB
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone F2C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human APOB.
Dilution ELISA Immunoprecipitation Radioimmunoassay The optimal working dilution should be determined by the end user.
Formulation In 0.01 M potassium phosphate, 2.5 mM KCl, 0.135 M NaCl (1 mg/mL BSA, 0.05% sodium azide).
Gene Accession No. P04114
Gene Alias FLDB
Gene Symbols APOB
Host Species Mouse
Immunogen Purified human serum LDL.
Purification Method Ion exchange chromatography purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338
Target Species Human
Content And Storage Store at 4°C for up to 6 months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.