Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ApoB, Mouse anti-Mouse, Rat, Hamster, Clone: 2G11, Millipore Sigma™

Catalog No. MABS204625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS204625 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS204625 Supplier MilliporeSigma Supplier No. MABS204625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detects Apolipoprotein B using this mouse monoclonal Anti-ApoB, clone 2G11 , Cat. No. MABS2046, is a mouse monoclonal antibody that detects Apolipoprotein B in Mouse, Rat, and Hamster and is tested for use in ELISA and Western Blotting.

Apolipoprotein B-100 (UniProt: E9Q414; also known as Apo B-100) is encoded by the Apob gene (Gene ID: 238055) in murine species. Apolipoprotein B is a major protein constituent of chylomicrons. Apo B particles are made up of lipids and proteins and each Apo B is bordered by a phospholipid monolayer with an inner core composed of variable amounts of triglycerides and cholesterol esters. Apo B is synthesized with a signal peptide (aa 1-27) that is subsequently cleaved off. Two isoforms of Apo B are produced- Apo B-100 is synthesized in the liver and Apo B-48 is produced in the intestine. Apo B-100 contains the region that binds to the LDL receptor. It contains VLDL particles, VLDL remnant particles and LDL particles. On the other hand, the Apo B-48 particles are made of chylomicrons and chylomicron remnant particles. Generally, LDL particles account for approximately 90% of Apo B particles in plasma. Abnormal trapping of an Apo B is a major factor in the development of atherosclerotic lesions. When Apo B particles degrade cholesterol is retained with macrophages that induce inflammatory response. Further entry and entrapment of Apo B can promote cardiovascular pathology over time. (Ref.: Sniderman, A et al. (2010). Nat. Rev. Endocrinol. 6(6); 335-346).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ApoB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G11
Conjugate Unconjugated
Dilution Western Blotting Analysis: 4 μg/mL from a representative lot detected ApoB in 10μg of rat liver and mouse liver tissue lysate. Western Blotting Analysis: A representative lot detected ApoB in Western Blotting applications (Nguyen, A.T., et. al. (2006). Biochem Biophys Acta. 1761(2):182-5; Cheng, D., et. al. (2016). J Biol Chem. 291(45):23793-23803). ELISA Analysis: A representative lot detected ApoB in ELISA applications (Nguyen, A.T., et. al. (2006). Biochem Biophys Acta. 1761(2):182-5).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Apolipoprotein B-100
Gene Symbols Apob
Host Species Mouse
Immunogen ApoB-containing lipoproteins isolated from Apoe -/- Apob 48/48 mice.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 2G11 detects Apolipoprotein B in Mouse, Rat, and Hamster plasma and tissues. It binds to ApoB-48 and ApoB-100, but not to ApoB-39.
Target Species Mouse, Rat, Hamster
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.