Learn More
Invitrogen™ APOB48R Monoclonal Antibody (OTI2E6)

Description
APOB48R Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | APOB48R |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI2E6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | APOBR |
| Gene Accession No. | Q0VD83 |
| Gene Alias | APOB100R; Apob48r; apoB-48R; Apobr; Apolipoprotein B receptor; apolipoprotein B100 receptor; apolipoprotein B-100 receptor; apolipoprotein B48 receptor; Apolipoprotein B-48 receptor; RGD1565169 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.