Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOBEC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519014
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519014 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519014 Supplier Invitrogen™ Supplier No. PA519014
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

This gene encodes a member of the cytidine deaminase enzyme family. The encoded protein forms a multiple-protein editing holoenzyme with APOBEC1 complementation factor and APOBEC1 stimulating protein . This holoenzyme is involved in the editing of C-to-U nucleotide bases in apolipoprotein B and neurofibromatosis-1 mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOBEC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene APOBEC1
Gene Accession No. P51908
Gene Alias Apobec1; APOBEC-1; apolipoprotein B editing complex 1; apolipoprotein B mRNA editing enzyme catalytic polypeptide 1; apolipoprotein B mRNA editing enzyme catalytic subunit 1; apolipoprotein B mRNA editing enzyme complex-1; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1; apolipoprotein B mRNA-editing enzyme 1; BEDP; C->U-editing enzyme APOBEC-1; Cdar1; HEPR; mRNA(cytosine(6666)) deaminase 1
Gene Symbols APOBEC1
Host Species Goat
Immunogen Synthetic peptide sequence (DVFYDPRELRKEAC) corresponding to the internal amino acids of APOBEC1 (aa 24-37).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11810
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.