Learn More
Invitrogen™ APOBEC2 Polyclonal Antibody

Description
Immunogen sequence: DDPEKLKELI ELPPFEIVTG ERLPANFFKF QFRNVEYSSG RNKTFLCYVV EAQGKGGQVQ ASRGYLEDEH AAAHAEEAFF NTILPAFDPA LRYNVTWYVS SSPCAACADR IIKTLSKTKN LRLLI Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 90%.
Specifications
Specifications
| Antigen | APOBEC2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | APOBEC2 |
| Gene Accession No. | Q9Y235 |
| Gene Alias | Apobec2; apolipoprotein B editing complex 2; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 2; apolipoprotein B mRNA editing enzyme catalytic subunit 2; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 2; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 2; ARCD1; Arp1; C->U-editing enzyme APOBEC-2; mRNA(cytosine(6666)) deaminase 2; OTTHUMP00000039773; probable C->U-editing enzyme APOBEC-2 |
| Gene Symbols | APOBEC2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.