Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOBEC3A/APOBEC3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104035
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104035 Supplier Invitrogen™ Supplier No. PA5104035
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total APOBEC3A/B.

PHO3 is a member of the cytidine deaminase gene family. The PHO3 gene is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOBEC3A/APOBEC3B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APOBEC3A
Gene Accession No. P31941, Q9UH17
Gene Alias A3A; A3B; APOBEC1L; APOBEC3A; APOBEC3B; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3A; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3B; apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3B; apolipoprotein B mRNA editing enzyme catalytic subunit 3A; apolipoprotein B mRNA editing enzyme catalytic subunit 3B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B; ARCD3; ARP3; ARP4; bK150C 2.2; bK150C2.1; bK150C2.2; cytidine deaminase; DJ742C19.2; DNA dC->dU-editing enzyme APOBEC-3A; DNA dC->dU-editing enzyme APOBEC-3B; FLJ21201; phorbolin 1; phorbolin 2; phorbolin 3; phorbolin-1; Phorbolin-1-related protein; phorbolin-2/3; PHRBN; PHRBNL; probable DNA dC->dU-editing enzyme APOBEC-3A; probable DNA dC->dU-editing enzyme APOBEC-3B
Gene Symbols APOBEC3A, APOBEC3B
Host Species Rabbit
Immunogen A synthesized peptide derived from human APOBEC3A(Accession P31941), corresponding to amino acid residues I26-L76.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200315, 9582
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.