Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APOBEC3A/APOBEC3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568281
Description
APOBEC3A/APOBEC3B Polyclonal Antibody for Western Blot
PHO3 is a member of the cytidine deaminase gene family. The PHO3 gene is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control.
Specifications
APOBEC3A/APOBEC3B | |
Polyclonal | |
Unconjugated | |
APOBEC3A | |
A3A; A3B; APOBEC1L; APOBEC3A; APOBEC3B; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3A; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3B; apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3B; apolipoprotein B mRNA editing enzyme catalytic subunit 3A; apolipoprotein B mRNA editing enzyme catalytic subunit 3B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B; ARCD3; ARP3; ARP4; bK150C 2.2; bK150C2.1; bK150C2.2; cytidine deaminase; DJ742C19.2; DNA dC->dU-editing enzyme APOBEC-3A; DNA dC->dU-editing enzyme APOBEC-3B; FLJ21201; phorbolin 1; phorbolin 2; phorbolin 3; phorbolin-1; Phorbolin-1-related protein; phorbolin-2/3; PHRBN; PHRBNL; probable DNA dC->dU-editing enzyme APOBEC-3A; probable DNA dC->dU-editing enzyme APOBEC-3B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
200315, 9582 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P31941, Q9UH17 | |
APOBEC3A, APOBEC3B | |
Synthesized peptide derived from human APOBEC3A/APOBEC3B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction