Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOBEC3B Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299174APC
Description
APOBEC3B Polyclonal antibody specifically detects APOBEC3B in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
APOBEC3B | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
APOBEC1L, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B, ARCD3, ARP4, bK150C2.2, cytidine deaminase, DJ742C19.2, EC 3.5.4, EC 3.5.4.-, FLJ21201, phorbolin 3, Phorbolin-1-related protein, phorbolin-2/3, PHRBNLphorbolin 2, probable DNA dC->dU-editing enzyme APOBEC-3B | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human APOBEC3B. | |
0.1 mL | |
Immunology | |
9582 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction