Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOBEC3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566863 Supplier Invitrogen™ Supplier No. PA566863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELRFLDLVPSL QLDPAQIYRV TWFI Highest antigen sequence identity to the following orthologs - mouse 52%, rat 52%.

This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. A hybrid gene results from the deletion of approximately 29.5 kb of sequence between this gene, APOBEC3B, and the adjacent gene APOBEC3A. The breakpoints of the deletion are within the two genes, so the deletion allele is predicted to have the promoter and coding region of APOBEC3A, but the 3' UTR of APOBEC3B. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOBEC3B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APOBEC3B
Gene Accession No. Q9UH17
Gene Alias A3B; A3Z2_1; A3Z2-Z3; A3Z3; A3Z3_1; APOBEC1L; Apobec3; Apobec3b; Apobec3f; APOBEC3F1; APOBEC3F2; APOBEC3Z2; APOBEC3Z3; apolipoprotein B editing complex 3; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3B; apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3B; apolipoprotein B mRNA editing enzyme catalytic subunit 3B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3F; apolipoprotein B mRNA-editing complex 3; ARCD3; Arp3; ARP4; BC003314; bK150C 2.2; bK150C2.2; Cem15; CEM-15; cytidine deaminase; DJ742C19.2; DNA dC->dU-editing enzyme APOBEC3; DNA dC->dU-editing enzyme APOBEC-3; DNA dC->dU-editing enzyme APOBEC-3B; FLJ21201; phorbolin 2; phorbolin 3; phorbolin-1-related protein; Phorbolin-2/3; PHRBNL; probable DNA dC->dU-editing enzyme APOBEC3; probable DNA dC->dU-editing enzyme APOBEC-3B; recovery from Friend virus 3; Rfv3
Gene Symbols APOBEC3B
Host Species Rabbit
Immunogen Recombinant Human APOBEC3B. Recombinant protein control fragment (Product #RP-105404).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9582
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.