Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOBEC3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121220
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121220 Supplier Invitrogen™ Supplier No. PA5121220
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, U-251MG, MCF7. The target is usually found in the following locations: Nucleus. Immunogen sequence: EEFAYCWENF VYNEGQQFMP WYKFDENYAF LHRTLKEILR YLMDPDTFTF NFNNDPLVLR RRQTYLCYEV ERLDNGTWVL MDQHMGFLCN EAKNLLCGFY.

This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. A hybrid gene results from the deletion of approximately 29.5 kb of sequence between this gene, APOBEC3B, and the adjacent gene APOBEC3A. The breakpoints of the deletion are within the two genes, so the deletion allele is predicted to have the promoter and coding region of APOBEC3A, but the 3' UTR of APOBEC3B. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOBEC3B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene APOBEC3B
Gene Accession No. Q9UH17
Gene Alias A3B; A3Z2_1; A3Z2-Z3; A3Z3; A3Z3_1; APOBEC1L; Apobec3; Apobec3b; Apobec3f; APOBEC3F1; APOBEC3F2; APOBEC3Z2; APOBEC3Z3; apolipoprotein B editing complex 3; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3B; apolipoprotein B mRNA editing enzyme catalytic polypeptide-like 3B; apolipoprotein B mRNA editing enzyme catalytic subunit 3B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3F; apolipoprotein B mRNA-editing complex 3; ARCD3; Arp3; ARP4; BC003314; bK150C 2.2; bK150C2.2; Cem15; CEM-15; cytidine deaminase; DJ742C19.2; DNA dC->dU-editing enzyme APOBEC3; DNA dC->dU-editing enzyme APOBEC-3; DNA dC->dU-editing enzyme APOBEC-3B; FLJ21201; phorbolin 2; phorbolin 3; phorbolin-1-related protein; Phorbolin-2/3; PHRBNL; probable DNA dC->dU-editing enzyme APOBEC3; probable DNA dC->dU-editing enzyme APOBEC-3B; recovery from Friend virus 3; Rfv3
Gene Symbols APOBEC3B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 151-250 of human APOBEC3B (NP_0048915).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9582
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.