Learn More
Invitrogen™ APOBEC3C Monoclonal Antibody (3E6)

Description
Peptide Sequence: YTSWSPCPDC AGEVAEFLAR HSNVNLTIFT ARLYYFQYPC YQEGLRSLSQ EGVAVEIMDY EDFKYCWENF VYNDNEPFKP WKGLKTNFRL LKRRLRESLQ.
Specifications
Specifications
| Antigen | APOBEC3C |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 3E6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | APOBEC3C |
| Gene Accession No. | Q9NRW3 |
| Gene Alias | A3C; APOBEC1L; APOBEC1-like; APOBEC3C; apolipoprotein B editing enzyme catalytic polypeptide-like 3C; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3C; apolipoprotein B mRNA editing enzyme catalytic subunit 3C; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3C; ARDC2; ARDC4; ARP5; bK150C2.3; DNA dC->dU-editing enzyme APOBEC-3C; MGC19485; PBI; Phorbolin I; probable DNA dC->dU-editing enzyme APOBEC-3C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.